Learn More
Description
A unique tool for maximum flexibility.
ScriptQuant is a brand-new image analysis module for QuantCenter. It provides a shell for user-created image analysis scripts in Python for maximum flexibility for the research user, enabling them to run their own image analysis algorithms within the QuantCenter framework.
Key Features
- Arbitrary script editor (VS Code by Microsoft is recommended)
- The user-created script can be saved into a scenario and handled like one
- Fast visual feedback: write your code and see the results immediately
- ScriptQuant can be extended with external libraries (e.g. OpenCV, TensorFlow)
- Well-documented API and use case examples
- ScriptQuant may provide a gateway for 3rd party AI developers
- Quantification results can be fed back into the 3DHISTECH infrastructure:
- Storage of quantification results (polygons, objects, regions, measured values)
- Visualization in SlideViewer as an overlay
- Statistics and Data visualization
- License for 5 users - Research use only
Specifications
Specifications
| Product Type | Image Analysis Script Module |
| For Use With (Application) | Digital Pathology |
| For Use With (Equipment) | Microarray Scanner |
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.