Learn More
Epredia™ 3DHistech SlideManager 20 Users, SlideCenter Slide 30 Users, and QuantServer 9 Users
Catalog No. 94423 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
Catalog No. For Use With (Equipment)
94-423 Microarray Scanner
1 options
Catalog No. 94-423 Supplier Epredia™ Supplier No. 94423
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech SlideManager, SlideCenter and QuantServer is a software package for integrated workflow management designed to support the researcher's work from defining the project through the collection and organization of samples to analysis and export.
SlideManager is an integrated workflow management software designed for research purposes, which supports the researcher's work from defining the project through the collection and organization of samples to analysis and export.
Key features of SlideManager:
  • Integrated and intuitive web-based user interface on which the entire workflow can be followed and managed.
  • The system enables the management of users and user roles, as well as the collaborative use of projects.
  • The system thinks in terms of research projects, which is a set of slides that can be hierarchized into groups. Metadata relevant to the research can be recorded for both the projects and the slides.
  • Integration with SlideCenter: from which users can import slides for the project.
  • The system enables the normalization of slides prepared and stained in different ways, after which the slides can be evaluated quantitatively due to the same coloring.
  • Integration with QuantServer: starting the normalization, region selection, image processing, evaluation, and display of data.
  • Integration with SlideViewer:
  • Integration SimpleSlideInterface: which allows researchers and external image processing software to access slides, folders, images, annotations, from any scripting or programming environment (e.g., Python script).
  • Backup and Restore features
  • License for 20 users.
SlideCenter is a flexible, distributed server system: more servers can easily be added for enhanced data security and storage requirements.
Licensing is performed on the server side: SlideCenter regulates the number of users who can access the server at any time. Five user types can be defined: Administrator user, Moderator user, Normal user, Restricted user and Public Share user.
Key Features
  • Automatic slide alignment
  • Virtual Slide and VirtualTray
  • TMA workflow support
  • Transparent login method for high-traffic user authentication
  • Server-side barcode parsing
  • InstantViewer
  • Web-based slide and case database with fast search
  • Easy extension by adding new SUs
  • Slide access through the free SlideViewer, the InstantViewer or the iPad Viewer
  • Virtual Slide handling
  • MS Network, HTTP and HTTPS accessibility
  • Fully manageable user and user group rights system
  • Direct scanning into database with PANNORAMIC digital slide scanners
  • Macro images and multiple file attachments stored in the case
  • SlideCenter can work with other 3DHISTECH software applications (e.g. quantification software can be run on remote slides)
  • License for 30 users.
  • QuantServer module allows you to execute processing and store the analyzation results on a dedicated server.
  • License for 9 users - Research use only.
Specifications
Product Type Software Package
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.