Learn More
Epredia™ 3DHistech SlideMaster Slide Format Conversion Solution - 1 Workstation
Catalog No. 91172 Shop All Epredia Products
Change view
Click to view available options
1 Workstation
Catalog No.
91-172 1 Workstation
1 options
Catalog No. 91-172 Supplier Epredia™ Supplier No. 91172
Add to Cart
Add to Cart
Request a Quote
The role of Epredia™ 3DHistech SlideMaster slide format conversion solution is to provide interoperability between different digital slide formats in whole-slide imaging.
SlideMaster provides a solution to convert digital slides from 3DHISTECH's proprietary MRXS slide format to other third-party slide formats, and vice versa.
Key Features
  • Directory monitoring and automated conversion
  • Annotation export
  • Upload and download with conversion
  • Flexible CPU core management
  • 1 workstation - Research use only
Specifications
Product Type Software License
For Use With (Equipment) Microarray Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.