Learn More
Epredia™ 3DHistech TMA Grand Master Automated Tissue Microarrayer, 72 Block Capacity
Catalog No. TMAGM Shop All Epredia Products
Change view
Click to view available options
For Use With (Application):
Digital Pathology
Catalog No. For Use With (Application)
TMAGM Digital Pathology
1 options
Catalog No. TMAGM Supplier Epredia™ Supplier No. TMAGM
Add to Cart
Add to Cart
Epredia™ 3DHistech TMA Grand Master is the fastest, highest-capacity and most user-friendly tissue microarrayer on the market.
An automated and sophisticated tissue microarrayer, allowing the precise selection and extraction of specific regions of interest on the donor blocks and their insertion in recipient blocks.
The TMA Grand Master elevates the tissue microarray workflow to a new level. With its high capacity, high speed and advanced features it is finally possible to create high-density and high-quality TMAs in a standardized way. Thanks to the clever hardware design, the block loading, block imaging, and arraying are parallel processes. Therefore, the TMA Grand Master can be loaded with new blocks continuously, even during the arraying process.
By using TMA Grand Master you can easily and quickly create TMA blocks with great precision, of 0.6, 1, 1.5 and 2 mm diameter tissue cores - Research use only.
Specifications
Product Type Automated Tissue Microarrayer
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.