Learn More
Epredia™ 3DHistech TMA Master II Automated Tissue Microarrayer, 5 Block Capacity
Catalog No. TMA2 Shop All Epredia Products
Change view
Click to view available options
For Use With (Application):
Digital Pathology
Catalog No. For Use With (Application)
TMA2 Digital Pathology
1 options
Catalog No. TMA2 Supplier Epredia™ Supplier No. TMA2
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech TMA Master II is the smallest fully automated tissue microarrayer on the market, it easily fits any laboratory bench.
  • TMA Master II is a robust and reliable computer-controlled instrument for creating tissue microarray (TMA) blocks.
  • It is an ideal solution for smaller laboratories, helping save reagent, slides, time, workforce and money.
  • By using TMA Master II, you can easily and quickly create TMA blocks with great precision, of 0.6, 1, 1.5 and 2 mm diameter tissue cores - Research use only.
Specifications
Product Type Automated Tissue Microarrayer
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.