Learn More
Medchemexpress LLC (3E,5E)-3,5-bis[(4-fluorophenyl)methylidene]-1-[(1-hydroxy-2,2,5,5-tetramethylpyrrol-3-yl)methyl] | 1172133-28-6 | 99.7% | 464.55 g/mol | C28H30F2N2O2 | 1 ML
SDP
Catalog No. 5000215113
Encompass_Preferred
HO-3867 is a synthetic curcumin analog that functions as a selective and potent STAT3 inhibitor with demonstrated antitumor activity in preclinical studies. It is supplied in ready-to-use solution and solid formats for laboratory research and should be handled according to standard safety procedures.
  • Selective STAT3 inhibitor with demonstrated antitumor activity.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexible use.
  • High purity, suitable for research applications.
  • Molecular weight approximately 464.55 g/mol.
  • Supplied for research use only; not for clinical use.
  • Requires low-temperature storage for long-term stability.
Catalog No. 50-002-15113 Supplier Medchemexpress LLC Supplier No. HY10045310MM 1ML
HO-3867 is a synthetic curcumin analog that functions as a selective and potent STAT3 inhibitor with demonstrated antitumor activity in preclinical studies. It is supplied in ready-to-use solution and solid formats for laboratory research and should be handled according to standard safety procedures.
  • Selective STAT3 inhibitor with demonstrated antitumor activity.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexible use.
  • High purity, suitable for research applications.
  • Molecular weight approximately 464.55 g/mol.
  • Supplied for research use only; not for clinical use.
  • Requires low-temperature storage for long-term stability.