Learn More
Cayman Chemical 3epI25hydroxy VItamIn D3 250ug
SDP
Catalog No. 502948532
Encompass_Preferred
The C-3 epimer of 25-hydroxy vitamin D3; decreases serum PTH levels in male, but not female, weanling rats
Catalog No. 50-294-8532 Supplier Cayman Chemical Supplier No. 34200250 UG
The C-3 epimer of 25-hydroxy vitamin D3; decreases serum PTH levels in male, but not female, weanling rats