Learn More
Cayman Chemical 3-Ethylmethcathinone (hydrochloride), Purity: ≥98%, CAS: 2446466-60-8, 10 mg
SDP
Catalog No. 502894391
Encompass_Preferred
A potential stimulant/entactogen designer drug of the amphetamine, phenethylamine, and cathinone chemical classes; intended for forensic purposes
Catalog No. 50-289-4391 Supplier Cayman Chemical Supplier No. 900108210 MG
A potential stimulant/entactogen designer drug of the amphetamine, phenethylamine, and cathinone chemical classes; intended for forensic purposes