Learn More
Cayman Chemical 3-Ethylmethcathinone (hydrochloride), Purity: ≥98%, CAS: 2446466-60-8, 50 mg
SDP
Catalog No. 502894388
Encompass_Preferred
A potential stimulant/entactogen designer drug of the amphetamine, phenethylamine, and cathinone chemical classes; intended for forensic purposes
Catalog No. 50-289-4388 Supplier Cayman Chemical Supplier No. 900108250 MG
A potential stimulant/entactogen designer drug of the amphetamine, phenethylamine, and cathinone chemical classes; intended for forensic purposes