Learn More
AdipoGen Bis(trifluoromethane)sulfonimide lithium salt
SDP
Catalog No. 501147146
Encompass_Preferred
Bis(trifluoromethane)sulfonimide lithium salt, AdipoGen Life Sciences. Chemical. CAS: 90076-65-6. Formula: C2F6LiNO4S2. MW: 287.09. Synthetic Used in the preparation of electrolyte suitable for lithium batteries showing very high conductivity. Used in the preparation of a chiral imidazolium salt via anion metathesis of the corresponding triflate.
Catalog No. 50-114-7146 Supplier AdipoGen Supplier No. CDXL0010G010
Bis(trifluoromethane)sulfonimide lithium salt, AdipoGen Life Sciences. Chemical. CAS: 90076-65-6. Formula: C2F6LiNO4S2. MW: 287.09. Synthetic Used in the preparation of electrolyte suitable for lithium batteries showing very high conductivity. Used in the preparation of a chiral imidazolium salt via anion metathesis of the corresponding triflate.