Learn More
Cayman Chemical 3-Fluoromethamphetamine (hydrochloride), Purity: ≥98%, CAS: 1324717-74-9, 50 mg
SDP
Catalog No. 502894344
Encompass_Preferred
An isomer of 4-FMA characterized by having fluorine at the 3 position of the phenyl group; intended for forensic and research applications
Catalog No. 50-289-4344 Supplier Cayman Chemical Supplier No. 900118550 MG
An isomer of 4-FMA characterized by having fluorine at the 3 position of the phenyl group; intended for forensic and research applications