Learn More
Cleatech Three Feet Bench top Vertical Laminar Flow station, Work Area 34"Wx 27"Dx 29.75"H ,
SDP
Catalog No. NC3136457
Encompass
Three Feet Bench top Vertical Laminar Flow station, Work Area 34"Wx 27"Dx 29.75"H ,Polypropylen & Powder coated Steel construction, Clear PVC front and side Windows (Sidepanels are upgradable to Polypropylene upon request), Back Plenum for air recirculation, Notincluding Fan/Filter Module and Worksurface (To be ordered separately). , Incl. Minihelic®gauge (0 - 1? Water Column)- (Choice of HEPA 99.99% efficient @ 0.3μm, or ULPA99.999% efficient @ 0.12μm 110VAC, 50/60Hz, Average airflow velocity 90 FPM ), IncludingBuilt-in LED Light
Catalog No. NC3136457 Supplier Cleatech Supplier No. 10002A
Three Feet Bench top Vertical Laminar Flow station, Work Area 34"Wx 27"Dx 29.75"H ,Polypropylen & Powder coated Steel construction, Clear PVC front and side Windows (Sidepanels are upgradable to Polypropylene upon request), Back Plenum for air recirculation, Notincluding Fan/Filter Module and Worksurface (To be ordered separately). , Incl. Minihelic®gauge (0 - 1? Water Column)- (Choice of HEPA 99.99% efficient @ 0.3μm, or ULPA99.999% efficient @ 0.12μm 110VAC, 50/60Hz, Average airflow velocity 90 FPM ), IncludingBuilt-in LED Light