Learn More
Avantor Fluid Handling C-Flex TPE Transfer Line, High Performance, Aseptic Welding, 3 ft
Catalog No. NC2961900
Encompass
C-Flex Thermoplastic Elastomer (TPE) tubing is ideal for biopharmaceutical and laboratory fluid processing. It offers exceptional tensile strength, chemical resistance, and low gas permeability, ensuring secure media transfer. This tubing supports various sterilization methods, is animal-derived component free, and is designed for aseptic connections, making it a reliable choice for single-use applications, tank transfers, and peristaltic pump use.
Catalog No. NC2961900 Supplier Avantor Fluid Handling Supplier No. CAS0276
C-Flex Thermoplastic Elastomer (TPE) tubing is ideal for biopharmaceutical and laboratory fluid processing. It offers exceptional tensile strength, chemical resistance, and low gas permeability, ensuring secure media transfer. This tubing supports various sterilization methods, is animal-derived component free, and is designed for aseptic connections, making it a reliable choice for single-use applications, tank transfers, and peristaltic pump use.