Learn More
Chemglass Life Sciences Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 3ft Length
SDP
Catalog No. 501531347
Encompass_Preferred
Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 3ft Length. Triple Insulated Flexible Hose. Flexible Stainless Steel tubing, insulated with fiberglass and silicone. Temperature Range -80 to 200C. One end has beaded pipe fitting and the other is a 3/4 Male NPT.
Catalog No. 50-153-1347 Supplier Chemglass Life Sciences Supplier No. CG1969B153
Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 3ft Length. Triple Insulated Flexible Hose. Flexible Stainless Steel tubing, insulated with fiberglass and silicone. Temperature Range -80 to 200C. One end has beaded pipe fitting and the other is a 3/4 Male NPT.