Learn More
Medchemexpress LLC 3H-1,2-benzodithiol-3-one-1,1-dioxide | 66304-01-6 | MFCD00132960 | 98.0% | 200.24 | C7H4O3S2 | 10 G
SDP
Catalog No. 5000212391
Encompass_Preferred
Beaucage reagent is an organosulfur compound used in nucleic acid chemistry as a DNA cleavage agent and as a sulfurizing reagent in oligonucleotide synthesis. It is supplied in both solid and solution formats for laboratory research use, with high purity suitable for analytical and preparative applications.
  • High purity suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • Useful for DNA cleavage and oligonucleotide sulfurization steps.
  • Provided in multiple pack sizes for flexibility in experiments.
  • Supplied with technical documentation and safety data for proper handling.
Catalog No. 50-002-12391 Supplier Medchemexpress LLC Supplier No. HY10095110G
Beaucage reagent is an organosulfur compound used in nucleic acid chemistry as a DNA cleavage agent and as a sulfurizing reagent in oligonucleotide synthesis. It is supplied in both solid and solution formats for laboratory research use, with high purity suitable for analytical and preparative applications.
  • High purity suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • Useful for DNA cleavage and oligonucleotide sulfurization steps.
  • Provided in multiple pack sizes for flexibility in experiments.
  • Supplied with technical documentation and safety data for proper handling.