Learn More
Medchemexpress LLC 3H-1,2-benzodithiol-3-one, 1,1-dioxide | 66304-01-6 | MFCD00132960 | 98.0% | 200.24 g·mol⁻¹ | C7H4O3S2 | 1 ML
SDP
Catalog No. 5000213362
Encompass_Preferred
Beaucage reagent is a small-molecule DNA-cleaving reagent used in nucleic acid research and oligonucleotide chemistry. It is commonly supplied as a 10 mM solution in DMSO for convenient handling in lab protocols. The material is documented at 98.0% purity and has a molecular weight of 200.24 g·mol⁻¹; follow supplier storage recommendations for optimal stability.
Catalog No. 50-002-13362 Supplier Medchemexpress LLC Supplier No. HY10095110MM 1ML
Beaucage reagent is a small-molecule DNA-cleaving reagent used in nucleic acid research and oligonucleotide chemistry. It is commonly supplied as a 10 mM solution in DMSO for convenient handling in lab protocols. The material is documented at 98.0% purity and has a molecular weight of 200.24 g·mol⁻¹; follow supplier storage recommendations for optimal stability.