Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-1-benzazepine-4-carboxylic acid, 2-amino-8-(1,1,2,2,2-pentafluoroethyl)-, ethyl ester | 926927-42-6 | MFCD11867857 | 99.4% | 348.267936 g·mol⁻¹ | C15H13F5N2O2 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1001810MM 1ML

Encompass_Preferred

Toll-like receptor modulator is a small-molecule research reagent reported to modulate TLR7 and TLR8 signaling and immune responses. Supplied as a light yellow to yellow powder with high reported purity, it is intended for in vitro and in vivo research, and the manufacturer provides stability, storage, and solvent guidance for preparation and dosing.

  • Modulates TLR7 and TLR8 signaling in immune research.
  • High reported purity (99.36% by LCMS).
  • Light yellow to yellow solid form suitable for formulation.
  • Recommended storage conditions for powder and solutions to maintain stability.
  • Soluble in DMSO for in vitro use and formulatable for in vivo dosing.
  • Available in small milligram pack sizes for research applications.

Catalog No. 50-002-15226


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.