Learn More
Medchemexpress LLC 3h-cyclopenta[c]quinoline, 4-(5-bromo-2-methoxyphenyl)-8-fluoro-3a,4,5,9b-tetrahydro | 415919-74-3 | 98.6% | C19H17BrFNO | 10MG
SDP
Catalog No. 5000259010
Encompass_Preferred
GPR30 agonist-1 is a selective small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER) for receptor pharmacology and vascular biology research. It exhibits concentration-dependent vasorelaxant effects in preclinical assays and is supplied with analytical documentation suitable for research use.
  • Small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER).
  • Reported vasorelaxant activity in preclinical studies.
  • High purity (98.6%) suitable for biochemical and cellular assays.
  • Available as a solid or as a 10 mM solution in DMSO for ready-to-use experiments.
  • SDS and certificate of analysis provided for quality verification.
  • Molecular formula C19H17BrFNO; molecular weight 374.25 g/mol.
Catalog No. 50-002-59010 Supplier Medchemexpress LLC Supplier No. HY13868610MG
GPR30 agonist-1 is a selective small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER) for receptor pharmacology and vascular biology research. It exhibits concentration-dependent vasorelaxant effects in preclinical assays and is supplied with analytical documentation suitable for research use.
  • Small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER).
  • Reported vasorelaxant activity in preclinical studies.
  • High purity (98.6%) suitable for biochemical and cellular assays.
  • Available as a solid or as a 10 mM solution in DMSO for ready-to-use experiments.
  • SDS and certificate of analysis provided for quality verification.
  • Molecular formula C19H17BrFNO; molecular weight 374.25 g/mol.