Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-cyclopenta[c]quinoline, 6,8-difluoro-3a,4,5,9b-tetrahydro-4-(3-pyridinyl)-, (3aS,4R,9bR)- | 1394285-50-7 | 99.5% | 284.30 g/mol | C17H14F2N2 | 100 MG
SDP

Catalog No. 5000213671
Encompass_Preferred

Golgicide A-2 (GCA-2) is a research chemical and the active enantiomer derivative of Golgicide A. It is used in cell biology and virology studies to probe ArfGEF GBF1-related pathways and antiviral activity. Supplied as a characterized solid for laboratory use.

  • High purity: 99.5%.
  • Cas number: 1394285-50-7.
  • Molecular formula: C17H14F2N2.
  • Molecular weight: 284.30 g/mol.
  • Physical form: solid, white to light brown.
  • Intended use: for research use only.
  • Pack size example: available in 100 mg quantities.

Catalog No. 50-002-13671 Supplier Medchemexpress LLC Supplier No. HY100540B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Golgicide A-2 (GCA-2) is a research chemical and the active enantiomer derivative of Golgicide A. It is used in cell biology and virology studies to probe ArfGEF GBF1-related pathways and antiviral activity. Supplied as a characterized solid for laboratory use.

  • High purity: 99.5%.
  • Cas number: 1394285-50-7.
  • Molecular formula: C17H14F2N2.
  • Molecular weight: 284.30 g/mol.
  • Physical form: solid, white to light brown.
  • Intended use: for research use only.
  • Pack size example: available in 100 mg quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.