Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-Imidazo[2,1-i]purine, 3-β-D-ribofuranosyl- | 39007-51-7 | 99.7% | C12H13N5O4 | 10MG
SDP

Catalog No. 5000259838
Encompass_Preferred

1,N6-Ethenoadenosine is a purine nucleoside analogue supplied as a research reagent for biochemical and nucleoside-analogue studies. It is provided as a solid with high chemical purity and defined solubility characteristics to support preparation of analytical and biological assays.

  • High purity: 99.7%.
  • Molecular formula C12H13N5O4; molecular weight 291.26 g/mol.
  • Soluble in water (125 mg/mL) and DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Supplied as a solid suitable for storage and handling.
  • Available in multiple package sizes including 10 mg.

Catalog No. 50-002-59838 Supplier Medchemexpress LLC Supplier No. HY13760010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,N6-Ethenoadenosine is a purine nucleoside analogue supplied as a research reagent for biochemical and nucleoside-analogue studies. It is provided as a solid with high chemical purity and defined solubility characteristics to support preparation of analytical and biological assays.

  • High purity: 99.7%.
  • Molecular formula C12H13N5O4; molecular weight 291.26 g/mol.
  • Soluble in water (125 mg/mL) and DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Supplied as a solid suitable for storage and handling.
  • Available in multiple package sizes including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.