Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-imidazo[4,5-b]pyridine, 2-ethyl-5,7-dimethyl- | 133240-06-9 | 99.4% | 175.24 g·mol^-1 | C10H13N3 | 25 MG
SDP

Catalog No. 5000208764
Encompass_Preferred

Adenosine receptor antagonist 4 is a small-molecule research compound that antagonizes the human adenosine A1 receptor, with a reported Ki of 101 nM. It is supplied as a solid and is intended for in vitro pharmacology and receptor-binding studies.

  • Acts as an adenosine A1 receptor antagonist (Ki ≈ 101 nM).
  • Molecular formula C10H13N3 and molecular weight ≈ 175.24 g·mol^-1.
  • High purity as reported in batch COA (≈99.35%).
  • Appearance: solid; recommended storage conditions vary by form (room temperature for solid; -80°C in solution).
  • Typical research-scale packaging: 25 mg vial (packaging inferred from supplier catalog number).

Catalog No. 50-002-08764 Supplier Medchemexpress LLC Supplier No. 25MGHYW063197
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine receptor antagonist 4 is a small-molecule research compound that antagonizes the human adenosine A1 receptor, with a reported Ki of 101 nM. It is supplied as a solid and is intended for in vitro pharmacology and receptor-binding studies.

  • Acts as an adenosine A1 receptor antagonist (Ki ≈ 101 nM).
  • Molecular formula C10H13N3 and molecular weight ≈ 175.24 g·mol^-1.
  • High purity as reported in batch COA (≈99.35%).
  • Appearance: solid; recommended storage conditions vary by form (room temperature for solid; -80°C in solution).
  • Typical research-scale packaging: 25 mg vial (packaging inferred from supplier catalog number).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.