Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-imidazo[4,5-f]quinoxalin-2-amine, 8-methyl-3-(methyl-d3) | 122457-31-2 | MFCD00210329 | 99.0% | 216.26 g/mol | C11H8D3N5 | 5 MG
SDP

Catalog No. 5000285110
Encompass_Preferred

MeIQx-d3 is the triply deuterated isotopologue of MeIQx supplied as a high-purity analytical standard for use as an internal standard and tracer in mass spectrometry and quantitative exposure studies.

  • Triply deuterated isotopologue for use as an internal standard.
  • High purity: 99.0%.
  • Molecular formula C11H8D3N5; molecular weight 216.26 g/mol.
  • Available as a 5 MG powder package.
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Applications include LC-MS/MS quantitation, tracer studies, and method development.

Catalog No. 50-002-85110 Supplier Medchemexpress LLC Supplier No. HYW355129S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MeIQx-d3 is the triply deuterated isotopologue of MeIQx supplied as a high-purity analytical standard for use as an internal standard and tracer in mass spectrometry and quantitative exposure studies.

  • Triply deuterated isotopologue for use as an internal standard.
  • High purity: 99.0%.
  • Molecular formula C11H8D3N5; molecular weight 216.26 g/mol.
  • Available as a 5 MG powder package.
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Applications include LC-MS/MS quantitation, tracer studies, and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.