Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3H-imidazo[4,5-f]quinoxalin-2-amine, 8-methyl-3-(methyl-d3)- | 122457-31-2 | MFCD00269884 | 99.0% | 216.26 g/mol | C11H8D3N5 | 1 MG
SDP

Catalog No. 5000285256
Encompass_Preferred

MeIQx-d3 is the deuterium-labeled analogue of MeIQx, provided for use as a tracer or internal standard in laboratory quantitative analysis. It is intended for research use only; MeIQx is a heterocyclic aromatic amine reported to be mutagenic and hepatocarcinogenic.

  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High chemical purity (99.0%).
  • Supplied in a small quantity suitable for trace-level analysis.
  • Molecular formula C11H8D3N5; molecular weight 216.26 g/mol.

Catalog No. 50-002-85256 Supplier Medchemexpress LLC Supplier No. HYW355129S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MeIQx-d3 is the deuterium-labeled analogue of MeIQx, provided for use as a tracer or internal standard in laboratory quantitative analysis. It is intended for research use only; MeIQx is a heterocyclic aromatic amine reported to be mutagenic and hepatocarcinogenic.

  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High chemical purity (99.0%).
  • Supplied in a small quantity suitable for trace-level analysis.
  • Molecular formula C11H8D3N5; molecular weight 216.26 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.