Learn More
Medchemexpress LLC 3H-indolium inner salt (TEA), N-hydroxysuccinimide ester | 1620475-28-6 | 95.0% | 1043.25 | C45H62N4O16S4 | 10 MG
SDP
Catalog No. 5000297462
Encompass_Preferred
Alexa Fluor 647 NHS ester (TEA) is a far-red, photostable succinimidyl ester dye supplied as the triethylammonium (TEA) salt for covalent labeling of primary amines on proteins, peptides, and amine-modified oligonucleotides. It produces strong fluorescence suitable for fluorescence imaging, flow cytometry, and other fluorescence-based assays. The product is supplied as a blue to dark blue solid and is sensitive to moisture and light.
  • Reactive succinimidyl ester for covalent labeling of primary amines.
  • Far-red fluorescence compatible with 633-647 nm excitation for imaging and flow cytometry.
  • Photostable with high signal intensity for prolonged imaging.
  • Suitable for labeling proteins, peptides, and amine-modified oligonucleotides.
  • Supplied as a blue to dark blue solid in TEA salt form; store sealed, away from moisture and light.
Catalog No. 50-002-97462 Supplier Medchemexpress LLC Supplier No. HYD2096A10MG
Alexa Fluor 647 NHS ester (TEA) is a far-red, photostable succinimidyl ester dye supplied as the triethylammonium (TEA) salt for covalent labeling of primary amines on proteins, peptides, and amine-modified oligonucleotides. It produces strong fluorescence suitable for fluorescence imaging, flow cytometry, and other fluorescence-based assays. The product is supplied as a blue to dark blue solid and is sensitive to moisture and light.
  • Reactive succinimidyl ester for covalent labeling of primary amines.
  • Far-red fluorescence compatible with 633-647 nm excitation for imaging and flow cytometry.
  • Photostable with high signal intensity for prolonged imaging.
  • Suitable for labeling proteins, peptides, and amine-modified oligonucleotides.
  • Supplied as a blue to dark blue solid in TEA salt form; store sealed, away from moisture and light.