Learn More
Medchemexpress LLC 3H-phenoxazin-3-one, 7-(acetyloxy)- | 1152-14-3 | MFCD00037659 | 98.0% | 255.23 | C14H9NO4 | 5 MG
SDP
Catalog No. 5000227489
Encompass_Preferred
Resorufin acetate is a fluorogenic resorufin ester used as a probe and substrate in biochemical and cell-based assays. It selectively detects hydrazine and is commonly used as a fluorometric substrate for esterases and aldehyde dehydrogenase activity; the compound is supplied as a light-sensitive white to yellow solid with reported purity of approximately 98.0%.
  • Fluorogenic substrate for esterase and aldehyde dehydrogenase activity
  • Sensitive probe for hydrazine detection
  • Produces a strong resorufin fluorescent signal upon enzymatic hydrolysis
  • Supplied as a light-sensitive solid; protect from light
  • High reported purity (~98.0%) suitable for analytical and cell-based assays
  • Recommended storage: 4°C; in solvent -80°C for long-term preservation
Catalog No. 50-002-27489 Supplier Medchemexpress LLC Supplier No. HY1182625MG
Resorufin acetate is a fluorogenic resorufin ester used as a probe and substrate in biochemical and cell-based assays. It selectively detects hydrazine and is commonly used as a fluorometric substrate for esterases and aldehyde dehydrogenase activity; the compound is supplied as a light-sensitive white to yellow solid with reported purity of approximately 98.0%.
  • Fluorogenic substrate for esterase and aldehyde dehydrogenase activity
  • Sensitive probe for hydrazine detection
  • Produces a strong resorufin fluorescent signal upon enzymatic hydrolysis
  • Supplied as a light-sensitive solid; protect from light
  • High reported purity (~98.0%) suitable for analytical and cell-based assays
  • Recommended storage: 4°C; in solvent -80°C for long-term preservation