Learn More
Medchemexpress LLC 3H-pyrrolo[2,3-d]pyrimidine-4,6-dione derivative (pentafluoropropoxy, trifluoroethoxy phenyl) | 1236767-85-3 | 99.9% | 473.27 | C17H11F8N3O4 | 50 MG
SDP
Catalog No. 5000228000
Encompass_Preferred
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor provided as a high-purity research compound for in vitro and in vivo studies. The compound shows low-nanomolar potency and includes solubility and formulation guidance in the datasheet and COA.
  • Selective delta-5 desaturase inhibitor with human IC50 22 nM and rat IC50 72 nM.
  • High reported purity: 99.9% (LCMS).
  • Molecular weight 473.27; formula C17H11F8N3O4.
  • Supplied as 50 MG research quantity.
  • DMSO solubility 125 mg/mL; in vivo formulation protocols provided.
  • Quality supported by an accompanying COA and datasheet.
Catalog No. 50-002-28000 Supplier Medchemexpress LLC Supplier No. HY11742750MG
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor provided as a high-purity research compound for in vitro and in vivo studies. The compound shows low-nanomolar potency and includes solubility and formulation guidance in the datasheet and COA.
  • Selective delta-5 desaturase inhibitor with human IC50 22 nM and rat IC50 72 nM.
  • High reported purity: 99.9% (LCMS).
  • Molecular weight 473.27; formula C17H11F8N3O4.
  • Supplied as 50 MG research quantity.
  • DMSO solubility 125 mg/mL; in vivo formulation protocols provided.
  • Quality supported by an accompanying COA and datasheet.