Learn More
Cayman Chemical 3hydroxy DecNIc ACD mthy 10mg
SDP
Catalog No. 502936025
Encompass_Preferred
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. putida and D. tsuruhatensis; has antibacterial activity against S. faecalis AD-4
Catalog No. 50-293-6025 Supplier Cayman Chemical Supplier No. 2461410 MG
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. putida and D. tsuruhatensis; has antibacterial activity against S. faecalis AD-4