Learn More
Cayman Chemical 3-hydroxy Desalkylgidazepam, Purity: ≥98%, CAS: 37891-18-2, 5 mg
SDP
Catalog No. 502943741
Encompass_Preferred
An analytical reference standard categorized as a benzodiazepine; an active metabolite of gidazepam and desalkylgidazepam; has been used as a precursor in the synthesis of substituted benzodiazepinones; intended for research and forensic applications
Catalog No. 50-294-3741 Supplier Cayman Chemical Supplier No. 422585 MG
An analytical reference standard categorized as a benzodiazepine; an active metabolite of gidazepam and desalkylgidazepam; has been used as a precursor in the synthesis of substituted benzodiazepinones; intended for research and forensic applications