Learn More
Cayman Chemical 3-hydroxy Lauric Acid methyl ester, Purity: ≥98%, CAS: 72864-23-4, 25 mg
SDP
Catalog No. 502936300
Encompass_Preferred
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. putida and D. tsuruhatensis
Catalog No. 50-293-6300 Supplier Cayman Chemical Supplier No. 2464325 MG
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. putida and D. tsuruhatensis