Learn More
Cayman Chemical 3-hydroxy Nonanoic Acid methyl ester, Purity: ≥98%, CAS: 83968-06-3, 25 mg
SDP
Catalog No. 503015786
Encompass_Preferred
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. oleovorans, P. putida, and D. tsuruhatensis
Catalog No. 50-301-5786 Supplier Cayman Chemical Supplier No. 2461225 MG
A hydroxy fatty acid methyl ester; has been found in PHA polymers produced by P. oleovorans, P. putida, and D. tsuruhatensis