Learn More
Cayman Chemical 3hydroxy OctNIc ACD mthy 10mg
SDP
Catalog No. 502934873
Encompass_Preferred
A hydroxy fatty acid methyl ester; a volatile compound and aromatic agent that has been found in pineapple, Yuzu, sudachi, and Kabosu sour citrus fruits, and human axillary sweat
Catalog No. 50-293-4873 Supplier Cayman Chemical Supplier No. 2461010 MG
A hydroxy fatty acid methyl ester; a volatile compound and aromatic agent that has been found in pineapple, Yuzu, sudachi, and Kabosu sour citrus fruits, and human axillary sweat