Learn More
Cayman Chemical 3-hydroxy Tridecanoic Acid methyl ester, Purity: ≥98%, CAS: 150024-70-7, 10 mg
SDP
Catalog No. 502932481
Encompass_Preferred
A hydroxy fatty acid methyl ester; major fatty acid component of A. lipolytica and A. glycerini; has been used in the synthesis of components of the female sex pheromone of the painted apple moth (T. anartoides)
Catalog No. 50-293-2481 Supplier Cayman Chemical Supplier No. 2464410 MG
A hydroxy fatty acid methyl ester; major fatty acid component of A. lipolytica and A. glycerini; has been used in the synthesis of components of the female sex pheromone of the painted apple moth (T. anartoides)