Learn More
Cayman Chemical 3-hydroxy-3-methylglutaryl-Coenzyme A (sodium salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502932729
Encompass_Preferred
An intermediate in several metabolic pathways; conversion to mevalonate by HMG-CoA reductase is the rate-limiting first step in the cholesterol biosynthetic pathway; can be cleaved into acetyl-CoA and the ketone body acetoacetate in mitochondria by HMG-CoA lyase; an intermediate in the degradation of leucine
Catalog No. 50-293-2729 Supplier Cayman Chemical Supplier No. 2539410 MG
An intermediate in several metabolic pathways; conversion to mevalonate by HMG-CoA reductase is the rate-limiting first step in the cholesterol biosynthetic pathway; can be cleaved into acetyl-CoA and the ketone body acetoacetate in mitochondria by HMG-CoA lyase; an intermediate in the degradation of leucine