Learn More
Cayman Chemical 3-hydroxy-4-Methoxyphenethylamine (hydrochloride), Purity: ≥98%, CAS: 645-33-0, 50 mg
SDP
Catalog No. 502911159
Encompass_Preferred
A metabolite of dopamine; decreases the viability of SH-SY5Y cells at 200 µM; induces tachycardia and increases blood pressure in normotensive or DOCA-salt hypertensive rats at 1 mg/kg; induces tremors and rigidity in mice at 100 mg/kg
Catalog No. 50-291-1159 Supplier Cayman Chemical Supplier No. 3601250 MG
A metabolite of dopamine; decreases the viability of SH-SY5Y cells at 200 µM; induces tachycardia and increases blood pressure in normotensive or DOCA-salt hypertensive rats at 1 mg/kg; induces tremors and rigidity in mice at 100 mg/kg