Learn More
Cayman Chemical 3-Hydroxy-4-methyl-2(5H)-thiophenone, Purity: ≥98%, CAS: 34876-35-2, 50 mg
SDP
Catalog No. 502912845
Encompass_Preferred
A degradation product of cefaclor; formed under aqueous acidic conditions
Catalog No. 50-291-2845 Supplier Cayman Chemical Supplier No. 3629050 MG
A degradation product of cefaclor; formed under aqueous acidic conditions