Learn More
Cayman Chemical 3hydroxy9 10DImethoxypTRo 10mg
SDP
Catalog No. 502911595
Encompass_Preferred
A flavonoid; inhibits PDGF-BB-induced proliferation of, and DNA synthesis in, A-10 vascular smooth muscle cells (IC50s = 10 and 1 µM, respectively)
Catalog No. 50-291-1595 Supplier Cayman Chemical Supplier No. 3615210 MG
A flavonoid; inhibits PDGF-BB-induced proliferation of, and DNA synthesis in, A-10 vascular smooth muscle cells (IC50s = 10 and 1 µM, respectively)