Learn More
Cayman Chemical 3HydroxyglutarIc ACD dImeth 1g
SDP
Catalog No. 502995177
Encompass_Preferred
A prochiral substrate; an esterified derivative of 3-hydroxyglutaric acid; used to characterize the ability of a lipase from C. antarctica to catalyze aminolysis of prochiral substrates
Catalog No. 50-299-5177 Supplier Cayman Chemical Supplier No. 229191 G
A prochiral substrate; an esterified derivative of 3-hydroxyglutaric acid; used to characterize the ability of a lipase from C. antarctica to catalyze aminolysis of prochiral substrates