Learn More
Cayman Chemical 3-Hydroxyglutaric Acid dimethyl ester, Purity: ≥95%, CAS: 7250-55-7, 25 g
SDP
Catalog No. 502994224
Encompass_Preferred
A prochiral substrate; an esterified derivative of 3-hydroxyglutaric acid; used to characterize the ability of a lipase from C. antarctica to catalyze aminolysis of prochiral substrates
Catalog No. 50-299-4224 Supplier Cayman Chemical Supplier No. 2291925 G
A prochiral substrate; an esterified derivative of 3-hydroxyglutaric acid; used to characterize the ability of a lipase from C. antarctica to catalyze aminolysis of prochiral substrates