Learn More
Cayman Chemical 3-Hydroxyisobutyrate (sodium salt), Purity: ≥95%, CAS: 1219589-99-7, 250 mg
SDP
Catalog No. 502936720
Encompass_Preferred
An intermediate in the metabolism of valine; increases fatty acid uptake by HUVECs at 0.1-100 mM; increases fatty acid uptake and lipid accumulation in mouse skeletal muscle; levels are increased in db/db diabetic mice and in humans with diabetes; human plasma levels are increased in obesity and correlate with a future risk of type 2 diabetes; human serum levels decrease following an exercise regimen of 2.5 hours of running for three days
Catalog No. 50-293-6720 Supplier Cayman Chemical Supplier No. 26105250 MG
An intermediate in the metabolism of valine; increases fatty acid uptake by HUVECs at 0.1-100 mM; increases fatty acid uptake and lipid accumulation in mouse skeletal muscle; levels are increased in db/db diabetic mice and in humans with diabetes; human plasma levels are increased in obesity and correlate with a future risk of type 2 diabetes; human serum levels decrease following an exercise regimen of 2.5 hours of running for three days