Learn More
Cayman Chemical 3-Hydroxypropionic Acid (sodium salt), Purity: ≥95%, CAS: 6487-38-3, 100 mg
SDP
Catalog No. 502937324
Encompass_Preferred
A building block in the synthesis of a variety of compounds with industrial applications, including acrylic acid, acrylamide, and 1,3-propanediol; accumulation is used as a biomarker of defects in propionic acid and branched-chain amino acid catabolism in patients with methylmalonic acidemia or propionic acidemia
Catalog No. 50-293-7324 Supplier Cayman Chemical Supplier No. 27603100 MG
A building block in the synthesis of a variety of compounds with industrial applications, including acrylic acid, acrylamide, and 1,3-propanediol; accumulation is used as a biomarker of defects in propionic acid and branched-chain amino acid catabolism in patients with methylmalonic acidemia or propionic acidemia