Learn More
Cayman Chemical 3hydroy PalmITC ACD mthy 10mg
SDP
Catalog No. 502984663
Encompass_Preferred
An esterized FA involved in quorum sensing; increases levels of PhcA-regulated virulence factors, greater than 20-, 30-, and 25-fold for EPS I, EGL, and PME, respectively, at a concentration of 175 nM
Catalog No. 50-298-4663 Supplier Cayman Chemical Supplier No. 2010810 MG
An esterized FA involved in quorum sensing; increases levels of PhcA-regulated virulence factors, greater than 20-, 30-, and 25-fold for EPS I, EGL, and PME, respectively, at a concentration of 175 nM