Learn More
Chemglass Life Sciences Coupling, 3" Beaded Pipe, "A" 4-1/4" X "B" 1-7/16", 75 in-lbs MAX Torque, 1/2" Hex Nut Size
SDP
Supplier:  Chemglass Life Sciences CG1968T75
Encompass_Preferred
Coupling, 3" Beaded Pipe, "A" 4-1/4" X "B" 1-7/16", 75 in-lbs MAX Torque, 1/2" Hex Nut Size. Complete coupling assembly consists of a 300 series Stainless Steel outer band with T-bolt, a Viton liner, and a PTFE sealing ring. Couplings allow for a maximum deflection of 4. PLEASE NOTE: TIGHTEN ALL COUPLINGS TO THE PROPER TORQUE SETTING.
Catalog No. 50-153-1330