Learn More
Cole-Parmer Ashcroft 20893.0 3" Digital Test Pressure Gauge 200 psi, 0.05% Accuracy
Catalog No. 50992541 Shop All Cole-Parmer Products
Encompass_Preferred
Gauge offers Total Error Band Accuracy (includes: terminal point, hysteresis, linearity, repeatability and temperature from 0/150F). Units of measure include: psi, inHg, inH2O, ftSW, bar, mbar, kPa, MPa, mmHg, cm2O, mm2O, kg/cm2. Seven selectable languages: English, Spanish, French, German, Italian, Portuguese, and Dutch
Catalog No. 50-992-541 Supplier Cole-Parmer Supplier No. 6843570
Gauge offers Total Error Band Accuracy (includes: terminal point, hysteresis, linearity, repeatability and temperature from 0/150F). Units of measure include: psi, inHg, inH2O, ftSW, bar, mbar, kPa, MPa, mmHg, cm2O, mm2O, kg/cm2. Seven selectable languages: English, Spanish, French, German, Italian, Portuguese, and Dutch