Learn More
Cole-Parmer Cole-Parmer 37130-9003 Egg-Shaped Stir Bar, 3" L x 3/4" Dia; 1/Pk
Catalog No. NC9408777 Shop All Cole-Parmer Products
Encompass_Preferred
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.
Catalog No. NC9408777 Supplier Cole-Parmer Supplier No. 0476885
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.