Learn More
Ace Glass, Inc. 3L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx185mm depth, with CAPFE O-ring, 28/15 O-ring ball joint inlet/outlet connects, pressure rated at 35 psi.
SDP
Catalog No. 50978673
Encompass
3L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx185mm depth, with CAPFE O-ring, 28/15 O-ring ball joint inlet/outlet connects, pressure rated at 35 psi.
Catalog No. 50-978-673 Supplier Ace Glass, Inc. Supplier No. 638624
3L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx185mm depth, with CAPFE O-ring, 28/15 O-ring ball joint inlet/outlet connects, pressure rated at 35 psi.