Learn More
Ace Glass, Inc. 3 liter epoxy coated flash chromatography reservoir with24/40 upper joint and 45/50 lower joint.
SDP
Catalog No. 50972461
Encompass_Preferred
Catalog No. 50-972-461 Supplier Ace Glass, Inc. Supplier No. 5871143