Learn More
Ace Glass, Inc. 3 liter epoxy coated flash chromatography reservoir with35/20 upper joint and 75/50 lower joint.
SDP
Catalog No. 50972494
Encompass_Preferred
3L epoxy coated flash chromatography reservoir with 35/20 upper joint and 75/50 lower joint. Reservoir connects between the column and flow control adapter.
Catalog No. 50-972-494 Supplier Ace Glass, Inc. Supplier No. 587248
3L epoxy coated flash chromatography reservoir with 35/20 upper joint and 75/50 lower joint. Reservoir connects between the column and flow control adapter.