Learn More
Ace Glass, Inc. Mantle, 3L, for 183mm diameter spherical or cylindrical flask, 183mm ID x 91.5mm height, 500w-115v, CSA rated
SDP
Catalog No. 50977325
Encompass
3L aluminum housed, table-model mantle for 183mm diameter spherical or cylindrical flask. 183mm I.D. x 91.5mm height. Maximum internal operating temperature is 450 Deg. centigrade. 500w-115v, cULus rated. Power cord included.
Catalog No. 50-977-325 Supplier Ace Glass, Inc. Supplier No. 1205323
3L aluminum housed, table-model mantle for 183mm diameter spherical or cylindrical flask. 183mm I.D. x 91.5mm height. Maximum internal operating temperature is 450 Deg. centigrade. 500w-115v, cULus rated. Power cord included.