Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass CGN-2165 3L High-Efficiency Polycarbonate Erlenmeyer Flask, Sterile, Nest
SDP

Catalog No. 502733465
Encompass

The 3 L high-efficiency polycarbonate Erlenmeyer flask is designed for single-use bioprocess and cell culture workflows. It is supplied sterile and nested for aseptic handling, with clear external graduations for easy volume observation and medical-grade polycarbonate construction for impact resistance and thermal tolerance.

  • Material: polycarbonate.
  • Volume: 3 L.
  • Sterility: sterile; USP Class VI.
  • Packaging: nest; sterile bags, 4 per case.
  • Temperature tolerance: compatible with temperatures up to 121 °C.
  • Transparency and scale: clear external graduation scale for observation.
  • Endotoxin and assay status: non-pyrogenic; passes endotoxin and cell culture tests.

Catalog No. 50-273-3465 Supplier Chemglass Life Sciences Supplier No. CGN21653L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 3 L high-efficiency polycarbonate Erlenmeyer flask is designed for single-use bioprocess and cell culture workflows. It is supplied sterile and nested for aseptic handling, with clear external graduations for easy volume observation and medical-grade polycarbonate construction for impact resistance and thermal tolerance.

  • Material: polycarbonate.
  • Volume: 3 L.
  • Sterility: sterile; USP Class VI.
  • Packaging: nest; sterile bags, 4 per case.
  • Temperature tolerance: compatible with temperatures up to 121 °C.
  • Transparency and scale: clear external graduation scale for observation.
  • Endotoxin and assay status: non-pyrogenic; passes endotoxin and cell culture tests.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.