Learn More
Chemglass Life Sciences Nest 3 L Wide-Mouth Erlenmeyer Flask, High Efficiency, Polycarbonate, Sterile
SDP
Catalog No. 502733462
Encompass
High-efficiency 3 L Erlenmeyer flask made from medical-grade polycarbonate, sterilized by E-beam and compliant with USP Class VI. Designed for cell culture and bioprocess applications, it offers clear graduations, a wide mouth for easy access, and impact and oxidation resistance suitable for standard laboratory handling.
  • Made from medical-grade polycarbonate with high transparency.
  • Sterilized by E-beam; non-pyrogenic.
  • Wide mouth for easy filling and sampling.
  • Accurate external graduation scale for volume observation.
  • Withstands temperatures up to 121°C.
  • Supplied with a solid plug-seal cap and packaged sterile in bags (4 per case).
Catalog No. 50-273-3462 Supplier Chemglass Life Sciences Supplier No. CGN21653WSC
High-efficiency 3 L Erlenmeyer flask made from medical-grade polycarbonate, sterilized by E-beam and compliant with USP Class VI. Designed for cell culture and bioprocess applications, it offers clear graduations, a wide mouth for easy access, and impact and oxidation resistance suitable for standard laboratory handling.
  • Made from medical-grade polycarbonate with high transparency.
  • Sterilized by E-beam; non-pyrogenic.
  • Wide mouth for easy filling and sampling.
  • Accurate external graduation scale for volume observation.
  • Withstands temperatures up to 121°C.
  • Supplied with a solid plug-seal cap and packaged sterile in bags (4 per case).